Learn More
EVOS™ EVOS™ Vessel Holder/Adapter for Automated Stage, with retention, multi-well plates
Catalog No. 12563556
Change view
Click to view available options
Quantity:
1 Holder
Catalog No. Quantity
12-563-556 1 Holder
1 options
Catalog No. 12-563-556 Supplier EVOS™ Supplier No. AMEPVH022
Add to Cart
Add to Cart
Allows user to utilize different types of vessels for imaging
  • Fits microscope slide, cell culture flask or dish, or micro-well plate to stage of EVOS imaging systems for increased precision in sample alignment
  • Holds multi-well plates
  • Acts as adapter for standard vessel holders (Cat numbers AMEP-VH001 thru AMEP-VH018)
  • Specific for automated stage of EVOS FL Auto Imaging System
  • Has lever for added convenience
TRUSTED_SUSTAINABILITY
Specifications
Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format one, multi-well plate
Holds Multi-well Plate
Quantity 1 Holder
How should the EVOS vessel holders be stored?
Store the EVOS vessel holders flat and protected from any contact with heavy objects as they can be distorted. Once the holders are distorted and no longer lay flat on the stage, they cannot be used. Store them individually in their own lidded plastic containers or small sturdy boxes.
How should I clean my EVOS vessel holders?
Wipe the vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol or 4,000 ppm hydrogen peroxide (H2O2).
I have an EVOS vessel that does not fit into any of the vessel holders that you offer. What are my options?
We do offer an option for making custom vessel holders. Please contact your Sales Representative for initiating a custom order.
For Research Use Only. Not for use in diagnostic procedures.