Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVOS™ EVOS™ Vessel Holder for Automated Stage, one Falcon T-75 flask or standard micro-well plate

Catalog No. 12563611
Change view
Click to view available options
Quantity:
1 holder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-563-611 1 holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-563-611 Supplier EVOS™ Supplier No. AMEPVH054
Only null left
Add to Cart
Add to Cart

EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.

Product Features:
  • EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
  • EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
  • With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
TRUSTED_SUSTAINABILITY

Specifications

Type Imaging System Vessel Holder
Product Line EVOS
Description Vessel Holder for Automated Stage, one Falcon T-75 flask or standard micro-well plate
For Use With (Equipment) EVOS™ FL Auto Imaging System
Holds 1 BD-Falcon T-75 flask, 1 multi-well plate
Dimensions 259 mm x 204 mm
Quantity 1 holder
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.