Learn More
Invitrogen™ EVOS™ Vessel Holder, KOVA™ Glasstic™ slide 10
Catalog No. 12563557
Change view
Click to view available options
Quantity:
1 Holder
Catalog No. Quantity
12-563-557 1 Holder
1 options
Catalog No. 12-563-557 Supplier Invitrogen™ Supplier No. AMEPVH017
Add to Cart
Add to Cart
Allows user to utilize different types of vessels for imaging
  • Fits microscope slide, cell culture flask or dish, or micro-well plate to stage of EVOS imaging systems for increased precision in sample alignment
  • Accommodates one KOVA Glasstic Slide 10
  • Easy-to-change vessel holders and stage plates available for this instrument platform
TRUSTED_SUSTAINABILITY
Specifications
Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format 10 Slides
Holds 1 KOVA™ Glasstic™ Slide 10
Quantity 1 Holder
How should the EVOS vessel holders be stored?
Store the EVOS vessel holders flat and protected from any contact with heavy objects as they can be distorted. Once the holders are distorted and no longer lay flat on the stage, they cannot be used. Store them individually in their own lidded plastic containers or small sturdy boxes.
How should I clean my EVOS vessel holders?
Wipe the vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol or 4,000 ppm hydrogen peroxide (H2O2).
I have an EVOS vessel that does not fit into any of the vessel holders that you offer. What are my options?
We do offer an option for making custom vessel holders. Please contact your Sales Representative for initiating a custom order.
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.