Learn More
EVOS™ EVOS™ Vessel Holder, one Nunc™ T-25 flask
Catalog No. 12563499
Change view
Click to view available options
Quantity:
1 holder
Catalog No. Quantity
12-563-499 1 holder
1 options
Catalog No. 12-563-499 Supplier EVOS™ Supplier No. AMEPVH018
Add to Cart
Add to Cart
EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.
Product Features:
  • EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
  • EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
  • With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
TRUSTED_SUSTAINABILITY
Specifications
Type Imaging System Vessel Holder
Product Line EVOS
Description Vessel Holder, one Nunc T-25 flask
For Use With (Equipment) EVOS™ FL Auto Imaging System
Holds 1 Nunc T-25 flask
Quantity 1 holder
How should the EVOS vessel holders be stored?
Store the EVOS vessel holders flat and protected from any contact with heavy objects as they can be distorted. Once the holders are distorted and no longer lay flat on the stage, they cannot be used. Store them individually in their own lidded plastic containers or small sturdy boxes.
How should I clean my EVOS vessel holders?
Wipe the vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol or 4,000 ppm hydrogen peroxide (H2O2).
I have an EVOS vessel that does not fit into any of the vessel holders that you offer. What are my options?
We do offer an option for making custom vessel holders. Please contact your Sales Representative for initiating a custom order.
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.