Learn More
Invitrogen™ EVOS™ Vessel Holder, universal
Catalog No. 12563494
Change view
Click to view available options
Quantity:
1 Holder
Catalog No. Quantity
12-563-494 1 Holder
1 options
Catalog No. 12-563-494 Supplier Invitrogen™ Supplier No. AMEPVH009
Add to Cart
Add to Cart
For flasks, trays and petri dishes
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
TRUSTED_SUSTAINABILITY
Specifications
Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format Universal
Holds All Vessel Types (Universal)
Quantity 1 Holder
How should the EVOS vessel holders be stored?
Store the EVOS vessel holders flat and protected from any contact with heavy objects as they can be distorted. Once the holders are distorted and no longer lay flat on the stage, they cannot be used. Store them individually in their own lidded plastic containers or small sturdy boxes.
How should I clean my EVOS vessel holders?
Wipe the vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol or 4,000 ppm hydrogen peroxide (H2O2).
I have an EVOS vessel that does not fit into any of the vessel holders that you offer. What are my options?
We do offer an option for making custom vessel holders. Please contact your Sales Representative for initiating a custom order.
For Research Use Only. Not for use in diagnostic procedures.