Learn More
Invitrogen™ EWSR1 Recombinant Rabbit Monoclonal Antibody (JE49-57)
Shop All Thermo Scientific Products

Description
Positive Control: K562, A431, SK-BR-3, rat testis tissue, human thyroid gland tissue, human skin tissue, human breast cancer tissue, human pancreas tissue, mouse kidney tissue, mouse heart tissue.
Specifications
Specifications
| Antigen | EWSR1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | JE49-57 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | EWSR1 |
| Gene Accession No. | Q01844 |
| Gene Alias | AC002059.7; bK984G1.4; Ewing sarcoma breakpoint region 1; Ewing sarcoma breakpoint region 1 protein; Ewing sarcoma homolog; Ewings sarcoma EWS-Fli1 (type 1) oncogene; Ews; EWS oncogene; EWS RNA binding protein 1; EWS RNA-binding protein 1; EWS RNA-binding protein variant 6; EWS-FLI1; Ewsh; EWSR1; RNA-binding protein EWS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.