Learn More
BIOGEMS INTERNATIONAL INC EX 527 1mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 49898335MG
Encompass
EX 527 is a selective inhibitor of NAD-dependent deacetylase sirtuin-1 (SIRT1) to the exclusion of histone deacetylases (HDACs) and other sirtuins. It is reported that inhibiting SIRT1 is associated with increasing insulin sensitivity and effecting other metabolic regulatory functions. EX 527 was used to determine that the inactivation of SirT1 stimulates tumor cell proliferation.
Catalog No. 50-153-0164
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.