Learn More
StatLab EXCEL PLUS PREFILL 20ML 48/PK
SDP
Supplier:  StatLab FXEXP20CS
Encompass_Preferred
Excell Plus is a pleasant smelling, low-hazard tissue fixative that works just like 10% formalin. Prefilled specimen containers are pre-labeled, leak-tight, and filled to 50% of the container's volume with ExCell Plus Fixative.
Catalog No. NC9734928
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.