Description
The new Excelis™ MPX-20C color camera delivers excellent performance for routine microscopy imaging applications. A Sony Exmor™ back-illuminated CMOS sensor is at the core of the MPX-20C and features a rolling shutter for high resolution and great color at fast frame rates. The large 1 in. sensor delivers a massive 20 MP image. Lightning fast data transfer across a USB 3.0 cable ensures stunning live views of the specimen. The new CaptaVision+ software also brings a new dimension to imaging with real-time stitching, real-time intelligent flatfield correction, depth of field fusion, dynamic measurements, and much more!
- Excellent color and fast frame rates
- Real-time stitching (via CaptaVision+)
- Depth of field fusion (via CaptaVision+)
- Intelligent flatfield correction (via Captavision+)
- Sony Exmor™ IMX183 color CMOS sensor
- View full 20 MP live images at 15 fps (faster frame rates at lower resolution)
- New CaptaVision+ Imaging Software (included) delivers full camera control, advanced image processing, customizable user interface, static and dynamic measurements with layers, and more (requires PC or Mac, not included).
- Standard c-mount easily adapts to most microscopes; 1.0x camera adapter recommended.
- 1-year warranty
- In the box: Excelis™ MPX-20C Camera, USB 3.0 Cable, CaptaVision+ PC Imaging Software
Specifications
Specifications
| Type | USB Microscope Camera |
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |