Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ExCellerate Human T Cell Expansion Media, Xeno-Free
SDP

Catalog No. CCM030 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM030 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CCM030 Supplier R&D Systems™ Supplier No. CCM030

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Media for the ex vivo expansion and culture of human T cell lymphocytes under xeno-free conditions

  • Robust T cell expansion under serum-free and xeno-free conditions
  • Optimized for use with the Cloudz™ T Cell Activation Kit
  • Compatible with a variety of T cell activation methods
  • Supports T cell expansion from PBMCs or purified CD3+ T cells

Specifications

For Use With (Application) T-Cell Expansion
Product Line R&D Systems
Quantity 500 mL
Content And Storage Store the unopened product at 2 - 8°C. Do not use past expiration date.
Description Facilitates discovery and preclinical research for T cell immunoregulation and therapy, robust platforms for ex vivo expansion and maintenance of T cells, including specialized serum-free or xeno-free media
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.