Learn More
Description
Specifications
Specifications
| Product Line | R&D Systems |
| Quantity | 500 mL |
| Species | Human |
| Content And Storage | Store the unopened product at ≤ -20 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Store opened product up to 2 weeks at 2-8 °C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
