Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Excelsior™ AS Tissue Processor Accessories

Catalog No. 7411258 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Potassium Permanganate Filter
Random cassette basket
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
74-112-58 Vapor Filter (Charcoal)
A78410024 Air System Duct Adapter Kit
A78410026 Storage Bottle
A78410027 Waste Wax Drawer Kit
B35210007 Basket Carrying Container
A78410021 Random cassette basket
B99-906-12CS Potassium Permanganate Filter
A78410025 Organized Basket, 222 cassette
A82310038 Organized Basket, 300 cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 74-112-58 Supplier Epredia™ Supplier No. 7411258
Add to Cart
Edge
Add to Cart

Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.

For use with Shandon Excelsior ES Tissue Processor

Specifications

Product Type Vapor Filter (Charcoal)
For Use With (Equipment) Excelsior AS Tissue Processor
Quantity Cs. of 6 Filters
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.