Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Excelsior™ AS Tissue Processor Accessories

Catalog No. A78410027 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Potassium Permanganate Filter
Random cassette basket
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
A78410027 Waste Wax Drawer Kit
A78410024 Air System Duct Adapter Kit
A78410026 Storage Bottle
B35210007 Basket Carrying Container
A78410021 Random cassette basket
74-112-58 Vapor Filter (Charcoal)
B99-906-12CS Potassium Permanganate Filter
A78410025 Organized Basket, 222 cassette
A82310038 Organized Basket, 300 cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. A78410027 Supplier Epredia™ Supplier No. A78410027
Add to Cart
Add to Cart
This item is not returnable. View return policy

Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.

For use with Shandon Excelsior ES Tissue Processor

Specifications

Product Type Waste Wax Drawer Kit
For Use With (Equipment) Excelsior ES Tissue Processor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.