Learn More
Excelta™ Handi-Vac Mini-Vacuum Pickup Replacement Bulb
Catalog No. 17468524 Shop All Excelta Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Handi-Vac Vacuum Pickup Bulb Kit
Catalog No. For Use With (Equipment)
17-468-524 Handi-Vac Vacuum Pickup Bulb Kit
1 options
Catalog No. 17-468-524 Supplier Excelta™ Supplier No. PVHVB
Add to Cart
Add to Cart
Squeeze and relax action of bulb gives fast pick-up and release of part to be handled
  • The PV-HV Handi-Vac™ is a vacuum-bulb pick-up-tool that is simple and easy to use. When the vacuum bulb is squeezed, place the soft suction cup on the part to be handled and release the vacuum bulb. A second squeeze releases the part.
  • The Handi-Vac kit comes with the following (4) Buna-N Anti-Static (101 to 1012 ohms/sq) vacuum cup/probe assemblies: PVB-CB-18, PVB-CB-14, PVS-CB-18 and PVS-CB-38.
Specifications
Product Type Replacement Bulb
For Use With (Equipment) Handi-Vac Vacuum Pickup Bulb Kit