Learn More
Excelta™ Pocket Beeper Continuity Tester
Catalog No. 17467649 Shop All Excelta Corporation Products
Change view
Click to view available options
PB-1 Pocket beeper
Catalog No.
17-467-649 PB-1 Pocket beeper
1 options
Catalog No. 17-467-649 Supplier Excelta™ Supplier No. PB1
Add to Cart
Add to Cart
For quick discernment of point-to-point continuity in production and field service applications
  • As a transistor and diode tester, it analyzes in circduit junctions for go/no go normality
  • Indicates leakage, shorted or open conditions of transistors
  • Uses 9V alkaline battery
Specifications
Product Type Pocket Beeper
Voltage 9 V
Amperage 5 mA
For Use With (Application) Quick discernment of point to point continuity in production and field service applications