Learn More
Excelta™ Replacement Blade For Medical Tubing Flush Cutters
Catalog No. 18100978 Shop All Excelta Corporation Products
Change view
Click to view available options
Cut Type:
Shear
Catalog No. Cut Type
18-100-978 Shear
1 options
Catalog No. 18-100-978 Supplier Excelta™ Supplier No. 292214X
Add to Cart
Add to Cart
Accessory for 2922 tubing cutters
This medical tubing cutter gives a nice clean shear cut to a variety of soft tubing materials. They feature stainless steel replaceable blades. The 2922-14-X is the replacement blade.
Specifications
Blade Material Stainless Steel
Cut Type Shear