Learn More
Excelta™ Roto-Pic™ Vacuum Handpiece
Catalog No. 17468531 Shop All Excelta Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Vacuum System, Ultra-precision
Catalog No. For Use With (Equipment)
17-468-531 Vacuum System, Ultra-precision
1 options
Catalog No. 17-468-531 Supplier Excelta™ Supplier No. 2000A
Add to Cart
Add to Cart
Five star replacement handpiece used with Roto-Pic vacuum system
The 2000A is the Roto-pic™ replacement hand piece. The hand piece includes a slide switch for on-off and a rotary wheel to turn the very tip for perfect alignment of the part.
Specifications
Product Type Roto-pic Vacuum Hand Piece
For Use With (Equipment) Vacuum System, Ultra-precision
For Use With (Application) For vacuum pickup of chips and wafers
Includes Positive hold-and-release slide bar, 360deg. rotatable round nose wheel, replaceable tips and silicon rubber cups