Learn More
Excelta™ Roto-Pic™ Vacuum System Vacuum Cup Kit
Catalog No. 17468538 Shop All Excelta Corporation Products
Change view
Click to view available options
Material:
Silicone Rubber
Catalog No. Material
17-468-538 Silicone Rubber
1 options
Catalog No. 17-468-538 Supplier Excelta™ Supplier No. 2000WP
Add to Cart
Add to Cart
Silicone rubber cups have brass fittings
The 2000WP Roto-pic™ vacuum cup kit contains cups that are made of silicone rubber with brass fittings. It contains one each of the 2050 (3/8 in.), 2051 (1/2 in.), 2052 (1/4 in.), 2060 (Circo-Vac) and 2071 (Vetro-Vac w/high temp support).
Specifications
Product Type Roto-pic Vacuum Cup Kit
Material Silicone Rubber
Diameter (English) Inside 0.37 in., 0.5 in., 0.25 in.
Diameter (Metric) Inside 1 cm, 1.2 cm, 0.6 cm
Includes 0.37, 0.5, 0.25 in. Cups, circo vac cup, verto vac cup and brass fittings