Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Scribes, Double-pointed

Catalog No. 17467636 Shop All Excelta Corporation Products
Change view
Click to view available options
Length (Metric):
216 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
17-467-636 216 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-467-636 Supplier Excelta™ Supplier No. 477
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features one straight point and one bent or hooked point, both heat-treated and hardened

  • Points are mounted in a knurled finger grip handle
  • Stainless steel construction in.

Specifications

Product Type Scribe, Double-pointed
Material Stainless Steel
Quantity Each
Tip Style Straight, Hooked
Length (English) 8.5 in.
Length (Metric) 216 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.