Learn More
Excelta™ Serrated Hemostat
Catalog No. 17467663 Shop All Excelta Corporation Products
Change view
Click to view available options
Length (Metric):
12.7 cm
15.2 cm
15.24 cm
Product Type:
Locking Hemostat (Cleanroom Safe)
Tip Style:
Curved Serrated
Straight Serrated
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Product Type Tip Style
17-467-663 15.2 cm Locking Hemostat (Cleanroom Safe) Curved Serrated
17-467-657 12.7 cm Locking Hemostat (Cleanroom Safe) Straight Serrated
17-467-660 12.7 cm Locking Hemostat (Cleanroom Safe) Curved Serrated
16-050-108 15.24 cm Locking Hemostat (Cleanroom Safe) Straight Serrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-467-663 Supplier Excelta™ Supplier No. 38SE
Add to Cart
Add to Cart
This Serrated Hemostat is made of stainless steel.
Cleanroom safe.
  • Mfr. No. 35-PH, 36-PH, 37-PH and 38-PH have vinyl coated handles
  • Mfr. No. 36-SE, 37-SE and 38-SE are clean room safe
Specifications
Product Type Locking Hemostat (Cleanroom Safe)
Length (English) 6 in.
Length (Metric) 15.2 cm
Material Stainless Steel
Size Medium
Tip Style Curved Serrated