Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Smooth-Jaw Hemostat

Catalog No. 17467658 Shop All Excelta Corporation Products
Change view
Click to view available options
Length (Metric):
12.7 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
17-467-658 12.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-467-658 Supplier Excelta™ Supplier No. 35ASE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Smooth-Jaw Hemostat is made of stainless steel.

  • Straight stainless steel (AISI 440) locking hemostats
  • Cleanroom safe
  • Feature serrated jaws except for 35-A-SE which has smooth jaws

Specifications

Product Type Locking Hemostat
Length (English) 5 in.
Length (Metric) 12.7 cm
Material Stainless Steel
Size Medium
Tip Style Smooth Jaw
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.