Learn More
Excelta™ Stainless Steel 5 in. Scalpel Handle
Catalog No. 18100974 Shop All Excelta Corporation Products
Change view
Click to view available options
Product Type:
Scalpel Handle (General Precision)
Scalpel Handle (High Precision)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18-100-974 Scalpel Handle (General Precision)
16-050-103 Scalpel Handle (High Precision)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-100-974 Supplier Excelta™ Supplier No. 177SE
Add to Cart
Add to Cart
Stainless steel scalpels featuring outside finger serrations
  • Choose either high and general precision
  • Stainless steel
  • Cleanroom safe and autoclavable
Specifications
Blade Material Stainless Steel
Product Type Scalpel Handle (General Precision)
Length (English) 5 in.
Length (Metric) 12.7 cm
Autoclavable Autoclavable