Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ White Ceramic Tip Tweezer Replacement Tips

Catalog No. 17456163 Shop All Protective Industrial Products Inc. Products
Catalog No. 17-456-163 Supplier Excelta™ Supplier No. HSCTM348NVXS
Only null left
Add to Cart
Add to Cart

For use with Excelta White Ceramic-Tip Tweezers (see cat. no. 17456041)

  • Reusable Sitewear Top long raglan sleeve, placket front with interfacing, knit cuffs, bound neck, slits at bottom of side seams.
  • ESD Microdenier 99% Polyester 1% Carbon

Specifications

Size X-Small
Gender Unisex
Material Polyester, Carbon
Color Navy
Neck Style Gripper
Seam Type Serged
Sleeve Style Raglan Sleeve
Disposable Reusable
Latex Free Yes
No. of Pockets None
Product Type Long Sleeve Shirt
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.