Learn More
Metrohm USA EXCHANGE UNIT 806 - 10ML BURET
SDP
Supplier:  Metrohm USA 63026210
Encompass
Exchange unit with integrated data chip with 10 mL glass cylinder and light protection. PCTFE/PTFE flat stopcock, FEP tubing connection, anti-diffusion buret tip and standard amber glass reagent bottle.
Catalog No. NC0329409