Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Executive Windrowe Mesh Back Guest Chair with Grade A Seat
Shop All Fisherbrand Products

Click to view available options
Color:
Borscht
Carbon
Marine Blue
Sterling
Storm
Taupe
Description
- Features adjustable lumbar support
- Comfort molded seat
- Steel seat base
- Antifungal/antibacterial Grade A vinyl
- Great compliment to Fisherbrand Executive Windrowe Mesh Back Chair
- Additional antibacterial protections
- High end look and feel
- Variety of color options to suit any work space
Specifications
Specifications
| Product Type | General Purpose Seating |
| Weight (English) | 25 lb. |
| Color | Taupe |
| Seating Height | Desk Height |
| Base Style | Powder-coated steel base with non-skid pads |
| Base Material | Powder-coated Steel |
| Footrest | No |
| With Tilt | No |
| With Armrest | No |
| With Backrest | Yes |
| Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction