Learn More
Exel International Disposable Hypodermic Needles
Description
- Latex-free
- Sterile
- Nontoxic
- Nonpyrogenic
- Polypropylene hubs and boxes are color coded for easy gauge selection
Order Info
Each needle is individually packaged and ready for use
Specifications
Specifications
| Material | Polypropylene (Hub), Stainless Steel |
| Color | Brown |
| Needle Gauge | 19 G |
| Latex Free | Yes |
| Length (English) Needle | 1 in. |
| Product Type | Hypodermic Needle |
| Needle Point Style | Regular Bevel |
| Sterility | Sterile |