Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exel International Disposable Spinal Needles

Catalog No. 1484123
Change view
Click to view available options
Needle Gauge:
18 G
20 G
22 G
25 G
Color:
Black
Orange
Pink
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
14-841-23 18 G Pink
14-841-25 22 G Black
14-841-24 20 G Yellow
14-841-26 25 G Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-841-23 Supplier Exel International Supplier No. 26960
Add to Cart
Add to Cart
This item is not returnable. View return policy

  • Latex-free
  • Sterile
  • Nontoxic
  • Nonpyrogenic
  • Convenient blister package
  • Siliconized for smooth, comfortable penetration

Specifications

Color Pink
Needle Gauge 18 G
For Use With (Application) Siliconized for smooth, comfortable penetration
Latex Free Yes
Length (English) Needle 3.5 in.
Product Type Spinal Needle
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.