Learn More
Exel International Scalp Vein Butterfly Sets
Catalog No. 1484033
Change view
Click to view available options
Needle Gauge:
19 G
20 G
21 G
23 G
25 G
27 G
Color:
Brown
Dark Blue
Gray
Green
Light Blue
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
14-840-33 20 G Yellow
14-840-32 19 G Brown
14-840-34 21 G Green
14-840-35 23 G Light Blue
14-840-38 27 G Gray
14-840-37 25 G Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 14-840-33 Supplier Exel International Supplier No. 26703
Add to Cart
Add to Cart
This item is not returnable. View return policy
•    Latex free
•    Individually packaged
•    Thin, stainless steel cannula is siliconized for easy insertion
•    Sterilized with Ethylene Oxide and ready for immediate use
•    Flexible wings hold the needle in position and help to prevent inadvertent pull out
•    Color-coded wings for easier identification
•    Large diameter needle permits greater flow of fluids
•    12 in. tube length
Specifications
Product Type IV Infusion Set
Color Yellow
Needle Gauge 20 G
Sterility Sterile
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.