Learn More
Exel International Winged IV Butterfly Blood Draw Set
Catalog No. 1484104
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Color:
Dark Blue
Green
Light Blue
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
14-841-04 21 G Green
14-841-05 23 G Light Blue
14-841-06 25 G Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-841-04 Supplier Exel International Supplier No. 26764
Add to Cart
Add to Cart
This item is not returnable. View return policy
•    Individually packaged
•    Thin, stainless steel cannula, siliconized for easy insertion
•    Sterilized with Ethylene Oxide and ready for immediate use
•    Flexible wings hold the needle in position and help to prevent inadvertent pull out
•    Color-coded for easier identification
Specifications
Product Type Winged IV Blood Draw Set
Color Green
Needle Gauge 21 G
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.