Learn More
Selleck Chemical LLC Exenatide Acetate 50mg 914454-01-6 H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2
SDP
Supplier:  Selleck Chemical LLC P104650MG
Encompass_Preferred
Exenatide Acetate, a 39-amino acid peptide originally isolated from the salivary glands of the Gila monster (Heloderma suspectum), differs from exendin-3 only in two positions close to the N-terminus. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8044
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.