Learn More
CPC Scientific H-His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific EXED005A
Encompass
A new member of the Glucagon superfamily that was originally isolated from the venom of the lizard, Heloderma Horridum. At low concentrations, it interacts with putative exendin receptors causing an increase in pancreatic acinar cAMP, while at higher concentrations it interacts with VIP receptors to stimulate an increase in cellular cAMP and amylase release. ONE-LETTER SEQUENCE: HSDGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2MOLECULAR FORMULA: C184H282N50O61S1MOLECULAR WEIGHT:4202.63STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [130357-25-4], RESEARCH AREA: DiabetesREFERENCES: J. Eng et al., J. Biol. Chem., 267, 7402 (1992)
Catalog No. 50-210-9269
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.