Learn More
Bachem Exendin-4 (1-8) Trifluoroacetate
SDP
Supplier:  Bachem 4051740.0005
Encompass_Preferred
Exendin-4 (1-8) Trifluoroacetate, H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-NH2, 1802078-29-0, C35H51N11O13, Mr 833.86, 5mg.
Catalog No. 50-260-087