Learn More
CPC Scientific H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2 0.5MG
SDP
Supplier:  CPC Scientific EXED003A
Encompass
Exendin-4 (Exenatide), originally isolated from Heloderma suspectum venom, stimulates acinar cAMP without causing amylase release, binds with similar affinity to the glucagon-like peptide 1 receptor, and is used in the treatment of type 2 diabetes.SEQUENCE: H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2ONE-LETTER SEQUENCE: HGEGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2MOLECULAR FORMULA: C184H282N50O60S1MOLECULAR WEIGHT: 4186.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [141758-74-9]SYNONYMS: ExenatideRESEARCH AREA: DiabetesREFERENCES:J. Eng et al., J. Biol. Chem., 267(11), 7402 (1992)SKU(s): EXED-003A, EXED-003B
Catalog No. 50-210-9265
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.