Learn More
Invitrogen™ Exendin 4 Monoclonal Antibody (35)

Description
ABS 012-35-02 detects exendin-4 from human samples. This antibody does not cross react with GLP-1, GLP-2 or glucagon. ABS 012-35-02 has been successfully used in ELISA procedures. The binding between ABS 012-35-02 and exendin-4 is disrupted by 4.5 M MgCl2. ABS 012-35-02 binds to an epitope in the 9-39 amino acid region of extendin-4. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Specifications
Specifications
| Antigen | Exendin 4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 35 |
| Concentration | 1.02 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene Alias | Exenatide |
| Host Species | Mouse |
| Immunogen | Synthetic extendin-4. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.