Learn More
AnaSpec EXENDIN (9-39) 1MG
Supplier:  AnaSpec AS24468
Encompass
Exendin (9-39)[DLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2]; MW 3369.8; (1 mg) This truncated Exendin-4 peptide, Exendin (9-39) amide, is a potent Glucagon-Like Peptide 1 (GLP-1) receptor antagonist. Unlike the full length Exendin-4 (a GLP-1 agonist), Exendin (9-39) antagonizes GLP-1–stimulated insulin release after food intake. It is a competitive inhibitor of Exendin-3 and Exendin-4.
Catalog No. NC0797096