Learn More
Bachem Exendin (9-39) Acetate
SDP
Supplier:  Bachem 4017799.0500
Encompass_Preferred
Exendin (9-39) Acetate, H-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2, 133514-43-9, C149H234N40O47S, Mr 3369.80, 0.5mg. Exendin (9-39) is a potent glucagon-like peptide 1 (GLP-1) receptor antagonist. It has also been described as an antagonist of the putative exendin receptor. Exendin (9-39) blocks the stimulatory action of GLP-1 (7- 36) amide and of exendin-4 on cAMP production in pancreatic acini. Moreover, exendin (9-39) was shown to be safely used to abolish the incretin effect of GLP-1 without interfering with the control of insulin secretion by circulating nutrients.
Catalog No. 50-194-6885
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.