Learn More
Genscript Corporation EXENDIN (9-39) PEPTIDE
Supplier:  Genscript Corporation RP108720.5
Encompass_Preferred
Exendin (9-39) is an inverse agonist of the murine glucagon-like peptide-1 receptor. There are implications for basal intracellular cyclic adenosine 3', 5'-monophosphate levels and beta-cell glucose competence. Exendin (9-39) is a competitive inhibitor of Exendin-3 and Exendin-4. On digestive smooth muscle, exendin (9-39) behaves as an antagonist for two members of the glucagon-receptor family, GLP-1 and glicentin.
Catalog No. 50-274-987
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.