Specifications
| Certifications/Compliance | ANSI Z87.1 |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Silver, Mirror |
| Frame Color | Gloss Black |
| Weight (Metric) | 29 g |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |