Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-exo-6-Hydroxytropinone 98.0+%, TCI America™
SDP

Catalog No. H12891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12891G Supplier TCI America Supplier No. H12891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5932-53-6
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.197
MDL Number MFCD00135648
InChI Key UOHSTKWPZWFYTF-LSBSRIOGSA-N
Synonym 6-hydroxytropinone, 1r,5r-6-hydroxy-8-methyl-8-azabicyclo 3.2.1 octan-3-one, 6-hydroxy-tropinone, 1r,5r-6-hydroxy-8-methylazabicyclo 3.2.1 octan-3-one
PubChem CID 51346151
IUPAC Name (1R,5R)-6-hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-one
SMILES CN1C2CC(C1CC(=O)C2)O

Specifications

Melting Point 123°C
Quantity 1 g
Formula Weight 155.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-exo-6-Hydroxytropinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.