Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101332 Supplier Invitrogen™ Supplier No. PA5101332
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EXO1.

Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXO1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EXO1
Gene Accession No. Q9QZ11, Q9UQ84
Gene Alias 5730442G03Rik; Exo1; EXOI; Exonuclease 1; exonuclease I; HEX1; hExo1; hExoI; mExo1; Msa; rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1
Gene Symbols EXO1
Host Species Rabbit
Immunogen A synthesized peptide derived from human EXO1(Accession Q9UQ84), corresponding to amino acid residues L66-R116.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26909, 305000, 9156
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.