Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXO5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523980
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523980 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523980 Supplier Invitrogen™ Supplier No. PA523980
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EXO5 Polyclonal Antibody for Western Blot, Flow

Single-stranded DNA (ssDNA) bidirectional exonuclease involved in DNA repair. Probably involved in DNA repair following ultraviolet (UV) irradiation and interstrand cross-links (ICLs) damage. Has both 5'-3' and 3'-5' exonuclease activities with a strong preference for 5'-ends. Acts as a sliding exonuclease that loads at ssDNA ends and then slides along the ssDNA prior to cutting; however the sliding and the 3'-5' exonuclease activities are abolished upon binding to the replication protein A (RPA) complex that enforces 5'- directionality activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXO5
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Exo5
Gene Accession No. Q9CXP9, Q9H790
Gene Alias 3110037I16Rik; AV297100; C1orf176; defects in morphology 1 homolog; defects in morphology protein 1 homolog; Defects in morphology protein 1-like; Dem1; Exo V; EXO5; exonuclease 5; exonuclease V; hExo5; mExo5; probable exonuclease V
Gene Symbols Exo5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 166-193 amino acids from the central region of human DEM1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100753327, 64789, 73172
Target Species Human, Mouse, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.