Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOC3L2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983316
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3316 Supplier Bioss Supplier No. BS14660R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen EXOC3L2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EXOC3L2
Gene Accession No. Q2M3D2
Gene Alias 4933417E01Rik; AU042655; EXOC3L2; exocyst complex component 3 like 2; exocyst complex component 3-like 2; Exocyst complex component 3-like protein 2; HBV XAg-transactivated protein 7; HBV X-transactivated gene 7 protein; protein 7 transactivated by hepatitis B virus X antigen (HBxAg); XTP7
Gene Symbols EXOC3L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EXOC3L2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90332
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.