Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exonuclease III, 25000 U, G-Biosciences
SCI_ED

Catalog No. S001153
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001153 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001153 Supplier G-Biosciences Supplier No. 7862263
Add to Cart
Add to Cart

Exonuclease III catalyzes the sequential excision of mononucleotides from 3'-hydroxyl termini of duplex DNA, or the RNA strand of a DNA/RNA hybrid.

  • A limited number of nucleotides are removed during each binding event, resulting in coordinated progressive deletions within the population of DNA molecules.

Specifications

Packaging Vial
Enzyme Exonuclease
Sensitive to Heat Inactivation Yes
Product Type Exonuclease III
Research Category Genome Editing, Traditional Cloning, DNA Repair
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.