Learn More
                                    New England Biolabs, Inc. Exonuclease VII – 200 units
Supplier: New England Biolabs, Inc. M0379S
            Exonuclease VII, (Exo VII) derived from E. coli, cleaves single-stranded DNA (ssDNA) from both 5' to 3' and 3' to 5' direction. This enzyme is not active on linear or circular dsDNA. It is useful for removal of single stranded oligonucleotide primers from a completed PCR reaction when different primers are required for subsequent PCR reactions. Digestion of ssDNA by Exonuclease VII is metal-independent.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.