Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Exonuclease VII

Catalog No. 502576128
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-257-6128 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-257-6128 Supplier Thermo Scientific™ Supplier No. 70082Z1000UN
Add to Cart
Add to Cart

A strict single-strand directed enzyme with 5'→3' and 3'→5' exonuclease activities.

Exonuclease VII is a strict single-strand directed enzyme with 5'→3' and 3'→5' exonuclease activities, making it the only bi-directional exonuclease with single-stranded specificity. Exonuclease VII has no apparent requirement for divalent cations, as it is fully active in the presence of EDTA. Initial reaction products are acid insoluble oligonucleotides which are further hydrolyzed into acid soluble form. The products of limited digestions are small oligomers (dimers to dodecamers).

Properties:

  • Molecular Weight: xseA subunit = 51.8 kDa; xseB subunit = 8.8 kDa
  • Optimum Temperature: 37 °C
  • Optimum pH: 8.0
  • Inactivation: 95 °C for 10 min.

Purity:

Greater than 95% pure as determined by SDS-PAGE. Tested for contaminating endonucleases, double-stranded exonucleases, and ribonucleases.

Storage Buffer:

50 mM Tris-HCl (pH 8.0), 200 mM NaCl, 0.5 mM EDTA, 10 mM DTT, 50% glycerol.

Assay Conditions:

The reaction (50 μL) contains 50 mM Tris-HCl (pH 7.9), 50 mM potassium phosphate (pH 7.6), 8.3 mM EDTA, 10 mM 2-mercaptoethanol, denatured DNA, and enzyme.

Unit Definition:

One unit is the amount of enzyme required to catalyze the conversion of 1 nmol of nucleotide to acid-soluble nucleotide in 30 min at 37 °C under standard assay conditions.

Concentration:

10 units/μL

Source:

E. coli strain containing overproducing clones encoding both the large (xseA) and small (xseB) subunits of Exonuclease VII.

Protocol: Typical Reaction Conditions (50 μL)

  • 70 mM Tris-HCI, pH 8.0
  • 8 mM EDTA
  • 10 mM 2-mercaptoethanol
  • 1 μg DNA
  • 50 μg/mL
  • 0.2 units Exonuclease VII

Incubate at 37 °C for 30 min. Inactivate the enzyme by heating to 95 °C for 10 min. Note that Exonuclease VII is not inhibited by EDTA.

Note: A typical dilution buffer for use with Exonuclease VII is 50 mM Tris, pH 7.9, 1 mM DTT and 0.5 mg/mL acetylated BSA.

Applications:

  1. Mapping positions of introns in genomic DNA.
  2. Removal of vector DNA from inserts with poly (dA-T) tails.
  3. Removal of excess PCR primers.
  4. Removal of single-stranded over-hangs produced by restriction endonucleases.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Shipped on dry ice. Store at -20°C.
Enzyme Exonuclease
Compatible Buffer Storage Buffer
Quantity 1000 U
Product Type Exonuclease VII

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.