Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOSC1 Monoclonal Antibody (BM-3)
GREENER_CHOICE

Catalog No. 01674353
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-353 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-353 Supplier Invitrogen™ Supplier No. MA112584
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is not recommended for paraffin sections in immunohistochemistry applications. Only frozen sections should be used. A suggested positive control for this product is HL-60 cells.

This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC1
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BM-3
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene EXOSC1
Gene Accession No. Q9Y3B2
Gene Alias 2610035C18Rik; 2610104C07Rik; 2610312F07Rik; 3'-5' exoribonuclease CSL4 homolog; AI447561; CGI-108; CSL4; Csl4p; EXOSC1; exosomal core protein CSL4; exosome complex component CSL4; Exosome component 1; hCsl4p; homolog of yeast exosomal core protein CSL4; p13; RP11-452K12.9; SKI4; Ski4p
Gene Symbols EXOSC1
Host Species Mouse
Immunogen Nuclei from Pokeweed mitogen stimulated with human peripheral blood lymphocytes.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51013
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.