Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7207562 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1145 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1145 Supplier Proteintech Group Inc Supplier No. 167311AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.

Specifications

Antigen EXOSC10
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EXOSC10
Gene Accession No. P56960, Q01780
Gene Alias Autoantigen PM/Scl 2, EXOSC10, exosome component 10, p2, p3, p4, PM Scl, PM/Scl 100, PMSCL, PMSCL2, RRP6, Rrp6p
Gene Symbols Exosc10
Host Species Rabbit
Immunogen EXOSC10 Fusion Protein Ag10189
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50912, 5394
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.