Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOSC3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501566389
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6389 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6389 Supplier Bethyl Laboratories, Inc Supplier No. A303909AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

EXOSC3 is component of the exosome 3′->5′ exoribonuclease complex and is required for the 3′ processing of the 7S pre-RNA to the mature 5.8S rRNA.

Specifications

Antigen EXOSC3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene EXOSC3
Gene Accession No. Q7TQK4, Q9NQT5
Gene Alias RP11-3J10.8, RRP40, Rrp40p, bA3J10.7, hRrp-40, hRrp40p, p10, exosome complex component RRP40, exosome complex exonuclease RRP40, exosome component Rrp40, ribosomal RNA-processing protein 40
Gene Symbols Exosc3
Host Species Rabbit
Immunogen Between 225 and 275
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51010, 66362
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.