Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOSC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100245 Supplier Invitrogen™ Supplier No. PA5100245
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EXOSC8.

This gene encodes a 3'-5' exoribonuclease that specifically interacts with mRNAs containing AU-rich elements. The encoded protein is part of the exosome complex that is important for the degradation of numerous RNA species. A pseudogene of this gene is found on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EXOSC8
Gene Accession No. Q96B26, Q9D753
Gene Alias 2310032N20Rik; bA421P11.3; CBP-interacting protein 3; CIP3; EAP2; EXOSC8; Exosome complex component RRP43; exosome complex exonuclease RRP43; exosome component 8; mKIAA4013; OIP2; OIP-2; Opa interacting protein 2; Opa-interacting protein 2; p9; PCH1C; Ribosomal RNA-processing protein 43; RP11-421P11.3; RRP43; Rrp43p
Gene Symbols EXOSC8
Host Species Rabbit
Immunogen A synthesized peptide derived from human EXOSC8(Accession Q96B26), corresponding to amino acid residues K63-I113.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11340, 295050, 69639
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.