Learn More
Exosome Detection (Simple Western) Antibody Pack, Novus Biologicals™
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Antigen | Exosome Detection (Simple Western) |
| Applications | Flow Cytometry, Immunohistochemistry, Simple Western, Western Blot |
| Conjugate | Unconjugated |
| Formulation | See individual datasheets |
| Immunogen | See individual datasheets |
| Quantity | 6 Vials |
| Regulatory Status | RUO |
| Target Species | Human, Mouse, Rat |
| Content And Storage | Storage is content dependent. |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.